Order Entry
Japan
Orders LinkContactUsLinkComponent
Hero_sf_810.jpg
Cut resistant gloves
Wells Lamont Industrial specializes in cut resistant gloves with many different products. From fabric to palm dipped to metal mesh gloves we cover many different applications and levels of cut resistance.
Wells Lamont Cut Resistant Gloves | Avantor
Block1_sf_810.jpg
The M121 glove liner offers ANSI 4 cut resistance and is highly reusable with great dexterity. Made with antimicrobial fibers.
Block2_sf_810.jpg
This glove offers superior cut resistance and features a patented self tensioning cuff that is easier to put on and more hygenic.
Block3_sf_810.jpg
The Y9265 has a polyurethane palm for grip and offers ANSI A2 cut resistance.
Block5_sf_810.jpg
The Y9290 offers a solid grip and ANSI A3 cut resistance to protect your hands from liquids while offering superior protection.
Block6_sf_810.jpg
Silver Talon- ANSI A6 cut resistance
The Silver Talon is approved for food contact, low lint, and is a great glove to protect your hands from extreme hazards. Made in the USA.