Order Entry
Japan
Orders LinkContactUsLinkComponent
brush_image.png
Specialty Brushes for Labware and Process Equipment
Brushes are essential for cleaning small diameter items, process equipment, portable tanks and storage vessels. With a robust range of styles and sizes, includng flexible stainless steel or polypropylene cores, and diameters from 1/4 inch to 5 inches, Micronova brushes meet lab and production cleaning challenges.
Specialty Brushes for Labware and Process Equipment | Avantor
Bottle_Brush_blue_Micronova_200.jpg
Bottle Brush
Color-coded stiff polyester bristle brushes with polypropylene handles for process equipment and parts. BPA-free base resin, constructed of FDA approved materials.
Cylinder_Brush_Micronova_200.jpg
Dome shaped, lightweight yet durable polyester bristle brush. Attaches to threaded handles for extended reach. BPA-free base resin, constructed of FDA approved materials.
Kettle_Brush_Micronova_200.jpg
Kettle Brush
Oblong shaped, lightweight, and durable brush. Attaches to threaded handles. Polyester bristles, BPA-free base resin, constructed of FDA approved materials.
valvebrush.png
Valve Brush
Large diameter, durable polyester bristle brush with attached polypropylene handle. Excellent for bowl and small vessels. BPA-free base resin, constructed of FDA approved materials.
FlexBrush_with_cover_Micronova_200.jpg
FlexBrush Curved Brush
A hook-shaped nylon bristle brush with stainless steel core. Its flexible core allows the user to shape the brush to custom applications including overhead piping and equipment interiors. Attaches to threaded handles.
Tube_Brush_Micronova_200.jpg
Plastic Tube Brush
100% polypropylene tube brushes. Ideal for cleaning finely polished stainless steel parts or tubing without scratching. A flexible core that glides through angled piping and fittings.
Brush_St_Sl_Micronova_200.jpg
Stainless Steel Wire Brushes
Durable stainless steel wire brushes with nylon bristles. Available in diameters of 1/4 to 2 inches and lengths from 12 to 72 inches.
Drain_Brush_Micronova_200.jpg
Drain Brush
A stiff black bristle, replaceable brush head, and a threaded plastic handle specifically designed and labeled for cleaning floor drains.
Scrub_Brush_Micronova_200.jpg
An iron-shaped plastic block handle with a comfortable, curved grip that protects hands from metal edges. Designed to clean tight crevices.