Order Entry
Japan
ContactUsLinkComponent
GreinerBioOneSeparation955x590.jpg
Greiner Bio-One Separation
With Leucosep, Greiner Bio-One optimized density gradient centrifugation whilst making it user-friendly. Alongside this, OncoQuick® was developed to extend the spectrum of applications to deal specifically with oncological targets.
GBO--Leucosep_150.jpg
Leucosep™ Centrifuge Tubes
Leucosep Tubes 12 mL and 50 mL sizes. Designed for easy separation of human monocytes and lymphocytes from peripheral blood.
  • Easy to fill without cumbersome layering steps
  • Easy recovery of interphase
  • No re-contamination with unwanted red blood cells
  • PBMC isolation from whole blood in 15 min
  • Un-filled or pre-filled with Leucosep separation medium
Learn More about Greiner Bio-One products
Product selection
greiner_easystrainer_100.jpg
EASYstrainer™ cell strainers are filters for Cell suspensions and primary cell isolates. Sterile. Fits all standard 50ml tubes. Venting slot for fast filtration.
greiner_leucosep_tubes_90.jpg
Leucosep developed for optimal separation of lymphocytes and peripheral mononuclear cells from human whole blood and bone marrow. Key feature is the porous barrier incorporated in the centrifuge tube.
greiner_separation_90.jpg
OncoQuick® is a simple-to-use, rapid and efficient system for the enrichment of circulating tumor cells that are released into the blood by a solid epithelial tumor or malignant melanoma...