Order Entry
Japan
ContactUsLinkComponent
GreinerBio_950.jpg
Greiner Bio-One Liquid Handling
Pipette Tips are a key component in day-to-day life science research and need to be of a high quality to ensure confidence and consistency in pipetting results. Our tips are manufactured from high-grade polypropylene to give the optimum in performance and fit.
Product selection
Pipettes_400x495.jpg
CELLSTAR® offers a wide range of different serological pipettes. Negative graduation to increase volume capacity and provided with color codes according to international standards
Petri_dishes_400x495.jpg
Petri dishes with diameters of 35, 60, 94, 100 and 145 mm. Manufactured from highly transparent polystyrene (PS), they are heat-resistant up to 60 °C for use with hot agar. Depending on the application vented and non-vented design can be selected.
Sapphire_400x495.jpg
Sapphire standard pipette tips, standard filter tips, low retention pipette tips and low retention filter tips. Free of detectable DNase, RNase, human DNA and are non-pyrogenic
Tubes_400x495.jpg
Tubes made of Polystyrene (ideal for optical measurements), Polypropylene (Displays high thermal, mechanical) and chemical resistance and Polyethylene (characterized by high therma and chemical resistance)
Erlenmeyer_400x495.jpg
Erlenmeyer shaker flasks are ideal for suspension culture of mammalian cells, plants, microbes and microorganisms. The shaker flasks feature a patented 2-in-1 DuoCAP® which allows for flexibility between vented and non-vented applications.