Order Entry
Japan
Orders LinkContactUsLinkComponent
GreinerBioOneImmunology_950x595.jpg
Greiner Bio-One Immunology
ELISA (Enzyme-Linked Immunosorbent Assay) is probably the most widely used biochemical method in laboratory analysis and diagnostics. Analytes such as peptides, proteins, antibodies and hormones can be detected selectively in low concentrations among a multitude of other substances and be quantified. Additionally, ELISAs are rapid, sensitive, cost-effective and can be performed in a high-throughput manner. ELISA is used in a variety of different assay types (e.g. direct ELISA, indirect ELISA, sandwich ELISA, competitive ELISA).
Greiner Bio-One Immunology | Avantor
Product selection
Microplates_400x495.jpg
96 Well ELISA Microplates
A wide variety of microplates for diagnostics and immunological research with different surface treatments and well profiles. The microplate footprint is compatible with automated systems.
StripPlates_400.jpg
Strip format microplates offer greater flexibility in diagnostics. Individual strips can be removed from the support frame so that the number of tests to be performed can be adjusted to the number of samples
ImmunoTubes_400.jpg
Immuno tubes are often used for determining hormone levels. With a length of 75 mm and a diameter of 12 mm with and without a “star”, they are available in both MICROLON® 200 and MICROLON® 600 quality
TerasakiPlates_400.jpg
Terasaki platesare small-sized microassay plates commonly used for microbatch protein crystallography. Conical wells avoid the spread of droplets away from the center for optimised crystallisation under oil.