Order Entry
Japan
Orders LinkContactUsLinkComponent
DWK Lab Bottles | Avantor
Lab Bottles
DWK Life Sciences offers a range of premium lab bottles that reflect the diverse needs of our customers. Chemical bottles to reagent bottles, clear or amber glass, Type I, II, or III, borosilicate or soda-lime — the DWK Life Sciences portfolio contains all this and more, including plastics in a wide variety of shapes, sizes, colors, and properties.
Product selection