Vista Sterile Cleanroom Overboots
ISOF80761
New Product
- Taglia dell'abbigliamento:Universal
- Sterili:+
- Applicazioni per abiti:Cleanroom
- Colore:bianco
- Descrizione:Vista cleanroom overboots
- Classe di cleanroom:10/ISO 4; EU GMP (A)
The Isofield Vista sterile cleanroom overboots are 50 cm (19,5”) tall and aseptically folded.
Sterile, cleanroom overboots
- Extra-long design, 50 cm knee height
- Two ties for added security
- Anti-slip PU sole
- Aseptically folded
Providing essential protection for GMP Grade A/B cleanroom applications, these anti-static, low-linting overboots are constructed with a non-slip PU sole, two sets of ties for secure fit, and excellent barrier properties.
:
Overboots are packaged 1 pair per inner bag, 10 pairs per outer bag, 8 outer bags per carton, 80 pairs per carton.