Order Entry
Italy
ContactUsLinkComponent
Strisce reattive, potassio, Reflectoquant®, Supelco®
Strisce reattive, potassio, Reflectoquant®, Supelco®
Catalog # 1.16992.0001
Supplier:  Merck
CAS Number:  
Strisce reattive, potassio, Reflectoquant®, Supelco®
Catalog # 1.16992.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Metodo chimico:
    Dipicrilammina
  • Conf:
    50 Tests
  • Descrizione:
    Potassio
  • M-Clarity quality level:
    MQ100
  • Range di misura:
    250 - 1200 mg/l K
Specifications
About this item
Strisce per l'uso su RQflex 10, RQflex 10 Plus e RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly