Mechanical cell counter
Catalog # 609-1145
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Range di misura:0 − 9999
- Descrizione:Mechanical counter
- Environmentally Preferable:
- Conf:1
Specifications
About this item
This manual steel cell counter allows for a mechanical counting from 0 to 9999.
- Finger lever
Colour: Chrome