Sealing tape, light blocking, non-fluorescent, Nunc™
Catalog # 731-0750
Supplier: Thermo Fisher Scientific
Specifications
- Proprietà ottica:Light blocking
- Descrizione:Nastro sigillante, blocco della luce
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:NAR Certificate of Retirement
- Conf:50
Specifications
About this item
Vinyl film with pressure sensitive acrylate adhesive. For application to plate top or bottom, to protect light sensitive samples.
- Temperature stable from −40 to +80 °C
- Non-pierceable by pipette
Colour: black
External L×W: 123×79 mm