
Tirofiban 97%
471301000.
New Product
:
- Tipo di conf.:Glass bottle
- Conf:100 mg
- Seq:2440
- Appearance:white to almost white powder
- Infrared spectrum:Conforms
- HPLC:≥96 %
:
:440.60 g/mol
:GHS07
:Refrigerata
:144494-65-5
:C₂₂H₃₆N₂O₅S
:MFCD05237246