PbS core-type quantum dots, oleic acid coated 10 mg/ml in toluene, giallo, liquido
Catalog # SIAL905062-5ML
Supplier: Merck
CAS Number:
Specifications
- Conf:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance (Color):Green to yellow
- Appearance (Form):Liquid
- Lead component:Confirmed
- FAPbBr3 quantum dots:Confirmed
- Emission peak:525 - 535 nm
- Quantum yield:≥70%
- Full width half max (FWHM):≤30 nm
Specification Test Results
About this item
:
Flash Pt:7 °C
:239.27 g/mol
:GHS02
:GHS07
:GHS08
:1314-87-0