Anti-4E-BP1 (Thr37/Thr46) phosphate Rabbit Monoclonal Antibody [clone: '236B4]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary Antibodies
Antigen: 4E-BP1 (Thr37/Thr46) phosphate
Clonality: Monoclonal
Clone: '236B4
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat,Monkey,D. melanogasterm
Specifications
- Catalog No:
- CSTA2855S
- CSTA2855L
- Simbolo dell'antigene:
- 4E-BP1 (Thr37/Thr46) phosphate
- 4E-BP1 (Thr37/Thr46) phosphate
- Tipo anticorpo:
- Primary
- Primary
- Clonalità:
- Monoclonale
- Monoclonale
- Clone:
- 236B4
- 236B4
- Reattività:
- Monkey, Human, D. melanogasterm, Rat, Mouse
- Monkey, Human, D. melanogasterm, Rat, Mouse
- Ospite:
- Rabbit
- Rabbit
- Isotipo:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChimica:
- Yes
- Yes
- ImmunoFluorescenza:
- Yes
- Yes
- Citometria a flusso:
- Yes
- Yes
- Temperatura di conservazione:
- −20 °C
- −20 °C
- Temperatura di spedizione:
- Ambient
- Ambient
Specifications
Frequently Bought Together