Order Entry
Italy
ContactUsLinkComponent
Anti-EIF2S1/eIF2-alpha (Ser52) phosphate Rabbit Monoclonal Antibody [clone: 119A11]
Anti-EIF2S1/eIF2-alpha (Ser52) phosphate Rabbit Monoclonal Antibody [clone: 119A11]
Supplier:  Cell Signaling
Certificates
Anti-EIF2S1/eIF2-alpha (Ser52) phosphate Rabbit Monoclonal Antibody [clone: 119A11]
Supplier:  Cell Signaling

Specifications

  • Catalog No:
  • CSTA3597S
  • CSTA3597L
  • Simbolo dell'antigene:
  • EIF2S1/eIF2-alpha (Ser52) phosphate
  • EIF2S1/eIF2-alpha (Ser52) phosphate
  • Tipo anticorpo:
  • Primary
  • Primary
  • Clonalità:
  • Monoclonale
  • Monoclonale
  • Clone:
  • 119A11
  • 119A11
  • Reattività:
  • Monkey, Human, D. melanogasterm, Rat, Mouse
  • Monkey, Human, D. melanogasterm, Rat, Mouse
  • Ospite:
  • Rabbit
  • Rabbit
  • Isotipo:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChimica:
  • Yes
  • Yes
  • ImmunoPrecipitazione:
  • Yes
  • Yes
  • Sequenza:
  • GMLLSELsRRRIRSC
  • GMLLSELsRRRIRSC
  • Temperatura di conservazione:
  • −20 °C
  • −20 °C
  • Temperatura di spedizione:
  • Cold
  • Cold

Specifications

Frequently Bought Together