Aggressive Adhesive Multi-Purpose Polyester Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
Catalog # BRDY170859
Supplier: Brady
Specifications
- Colore:nero su bianco
- Dimensioni:38,10W mm × 7,62L m
- Materiale:Poliestere
- Modello:M5C-1500-422
- Conf:1
- Adesivo:Acrilico permanente
Specifications
About this item
Easily label components with the thermal transfer, glossy permanent polyester label material (B-422). Print almost anywhere with a durable Brady handheld label printer.
- Versatile - Designed for a wide range of applications such as labeling equipment, circuit boards, panel components and barcodes
- Tough - Constructed to withstand abrasions, extreme temperatures and solvents such as alcohol, oil and water
- Adhesive - Made with a permanent acrylic adhesive that sticks to rough, smooth and powder-coated flat surfaces
Ideal for asset tracking, barcode labelling, component and equipment labelling, data and telecommunications labelling, general identification, inventory and inspection labelling applications.
Gloss finish, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant, outdoor use B-422 labels are compatible with Brady printer models M510, BMP51, BMP53, and M511.
Certifications: UL Recognized to UL969, CSA to C22.2 No.0.15-95, RoHS Compliant to 2011/65/EU (Pass 2015/863).