Powered air respirator over-visors, Versaflo™
Catalog # MMMAM-967
Supplier: 3M
Specifications
- Descrizione:Headgear cover
- Conf:1
Specifications
About this item
The Versaflo™ Over-Visor, gold, IR/UV shade 5, M-967, is an over-visor to protect against UV/IR, metal sparks, and splashes.
- Gold IR/UV over-visor
- For use with Versaflo™ Over-visor Frame, M-961
Get IR and UV protection when needed with the Versaflo™ Over-visor, Gold, IR/UV Shade 5, M-967. This over-visor measures 500×250 mm and protects users from UV as well as IR. It’s designed for use with 3M™ Versaflo™ Over-visor Frame, M-961.