Avantor® Hichrom HI-1 HT, GC Columns
Supplier: Avantor
Certificates
About this item
General purpose GC-MS column Solvent impurities, PCBs, SimDist, drugs, natural gases, essential oils, semivolatiles, pesticides, phenols, etc.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HI86-10-010-10
- HI86-25-025-15
- HI86-25-025-30
- HI86-25-015-30
- HI86-32-010-30
- HI86-32-015-15
- HI86-32-010-10
- HI86-32-010-15
- HI86-32-025-30
- HI86-32-015-30
- HI86-32-025-15
- HIGCHI873201015G5
- HIGCHI865301515G5
- HIGCHI86-53-088-10
- HI86-10-010-5
- HI86-32-010-10G2
- HI86-53-010-30
- HI86-53-015-15
- HI86-53-010-10
- HI86-53-010-15
- HI86-53-025-15
- HI86-53-025-30
- HI86-53-015-30
- HI86-53-017-30
- HI86-25-010-30
- HI86-25-015-15
- HI86-25-010-15
- Phase name:
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- HI-1 HT
- Phase composition:
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - high temperature
- Polarity:
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Limite di temperatura:
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- Codice USP:
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38