Apron with sleeves, MICROGARD® 2000, model 214
Catalog # ANSEWH20B-00214-07
Supplier: Ansell
Specifications
- Taglia dell'abbigliamento:3XL
- Applicazioni per abiti:Medical,Clinical
- Colore:bianco
- Seq:06
- Conf:60
Specifications
About this item
These aprons are made from a microporous PE film with a non woven inner layer. The fabric provides low hazard liquid chemical repllence particle protection, protection from pesticides and a barrier to biological agents.
- Can be used as a hospital gown
- Rear Velcro fastening
- Elasticated wrists
- Bound seams