Specifications
- Descrizione:Respiratory system, S-200+
- Seq:01
- Conf:1
Specifications
About this item
The 3M™ S-200+ supplied air respirator system is a comfortable and versatile supplied air respirator system for use with the 3M™ 6000 and 3M™ 7000 Series half and full face masks. Ideal for applications like spray painting, glass fibre fabrication, powder coating, material handling.
- Engineered to deliver additional comfort and security the double action safety couplings help to minimise the risk of accidental disconnection during use
- Lightweight and balanced system with a constant flow of air across the face for a cooler and more comfortable wear
- Easy to maintain, spare parts are quick, easy and intuitive to replace
- S-200+ includes a Flow Check Bag allowing to conduct a simple pre-use check to ensure that the correct amount of air is being delivered
Tested and approved to EN 14594:2005: Meets the highest level of protection, Class 4B for continuous flow supplied air respirators when fitted to a full face mask. When used with a 3M™ recommended Full Face mask the product meets the requirements for a Class 4A/B device. Assessed in accordance with the requirements of the ATEX directive and in accordance with EN13463-1 for non-electrical equipment (Valid only when fitted with a 3M™ 308-00-72P anti-static compressed air supply tube). Suitable for use in IIB 2GD environments under certain conditions.