Kitting cart
Kitting cart, single bay with 2-brake and 2-swivel rubber castors, 660×574×1727 mm, 25,2 kg
Codice catalogo METOPT1C-5M
Nome del fornitore: Metro International
Vai a:
Product Details & Documents
Kitting carts are ideal for handling totes, PCB carriers, trays or other material handling containers.
- Non conductive
Specifiche
- Descrizione:Kitting cart, single bay with 2-brake and 2-swivel rubber castors
- L×P×A:660×574×1727 mm
- Peso:25,2 kg