Microplate sealing tape
Codice catalogo GENO786-422
Nome del fornitore: G-Biosciences
Specifiche
- Descrizione:Self-adhesive sealing tape, for 96- and 384-well plates, 100 sheets
- Conf:1 KIT
Specifiche
Informazioni su questo articolo
Acetate film with acrylic adhesive. This self-adhesive microplate sealing tape is suitable for sealing both 96-well and 384-well plates. The precut, pressure-senstive tape seals and reseals the surface of the plates during incubations to prevent evaporation and allow more vigorous mixing. The tape will reseal the plates 2 to 3 times.
- Non sterile
- Transparent and non pierceable
- Minimise evaporation and condensation
- For short and long term storage of plates
- Temperature range: −16 to +38 °C