- Descrizione:Escape hood, Parat® 7530, with ABEK CO P3 filter, Hard case
- Seq:01
- Conf:1
This combined fire and industrial escape hood was developed with users, placing the focus on the fastest possible escape. Optimised operation and wearing comfort, a robust housing and a tested ABEK CO P3 filter ensure protection from toxic industrial and fire-related gases, vapours and particles for at least 15 minutes.
- Hood in signal colour for high visibility
- Automatically adjusting internal head harness: No additional tighting of the hood is required
- Flexible textile neck seal
- Exhalation valve directly leads the exhaled air out of the half mask
- Wide field of vision thanks to a specially formed visor with anti-fog coating
- Filter is automatically relased from the packaging when opened
- Mask fits a wide variety of face sizes
When opening the packaging, the filter plug is automatically released from the filter. The filter is then deployed into operational position and the hood can be immediately donned. Thanks to the self-adjusting internal head harness, no additional adjustment is required. Users only have to open the packaging, remove and don the hood – and leave the danger zone.
Total service life: 16 years (provided the filter is exchanged after eight years)