Chlorine sensitive 1 test strips, Dosatest® (0 • 0,05 • 0,1 • 0,2 • 0,4 • 0,8 • 1,2 mg/L Cl₂) 1 * 50 items
Catalog # 92536.650
Supplier: VWR Chemicals
New Product
Chlorine sensitive 1 test strips, Dosatest® (0 • 0,05 • 0,1 • 0,2 • 0,4 • 0,8 • 1,2 mg/L Cl₂) 1 * 50 items
Catalog # 92536.650
Supplier: VWR Chemicals
New Product
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK