Order Entry
Italy
ContactUsLinkComponent
Proteus mirabilis WDCM 00023 Vitroids 15-80 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000232-10EA
Supplier:  Merck
New Product
Proteus mirabilis WDCM 00023 Vitroids 15-80 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000232-10EA
Supplier:  Merck