Order Entry
Italy
ContactUsLinkComponent
Escherichia coli WDCM 00012 Vitroids 80-130 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000123-10EA
Supplier:  Merck
New Product
Escherichia coli WDCM 00012 Vitroids 80-130 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000123-10EA
Supplier:  Merck