Order Entry
Italy
ContactUsLinkComponent
5 ampoule de 5 mL 3 components: 2-Methyl-2-butene [CAS:513-35-9] 0.4% Chloroform [CAS:67-66-3] 0.4% Tetrachloromethane [CAS:56-23-5] 0.2% in Dichloromethane CRM, ISO 17034 and ISO 17025,, Conservation 12 months.,, 2°C to 8°C,, 1 * 5 Ampoul
Catalog # CMPLRD1219075.5
New Product
5 ampoule de 5 mL 3 components: 2-Methyl-2-butene [CAS:513-35-9] 0.4% Chloroform [CAS:67-66-3] 0.4% Tetrachloromethane [CAS:56-23-5] 0.2% in Dichloromethane CRM, ISO 17034 and ISO 17025,, Conservation 12 months.,, 2°C to 8°C,, 1 * 5 Ampoul
Catalog # CMPLRD1219075.5