Order Entry
Italy
ContactUsLinkComponent
Muffle Furnace L15/14/B510 Tmax1400 with flap door and protective gas connection, over-temperature limiter and thermocouple hole in furnace door 1 * 1 items
  455-3404
New Product
Muffle Furnace L15/14/B510 Tmax1400 with flap door and protective gas connection, over-temperature limiter and thermocouple hole in furnace door 1 * 1 items
  455-3404