Order Entry
Italy
ContactUsLinkComponent
Muffle Furnace L5/14/C550 Tmax1400 with flap door and protective gas connection and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3321
Supplier:  NABERTHERM
New Product
Muffle Furnace L5/14/C550 Tmax1400 with flap door and protective gas connection and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3321
Supplier:  NABERTHERM