Order Entry
Italy
ContactUsLinkComponent
4' Logic Class II, Type A2 Biological Safety Cabinet with 8 sash opening, Service Fixture, UV Lamp, Vacu-Pass Portals and Base Stand, 230V, 50/60Hz, Schuko 1 * 1 items
Catalog # LBCP302481132
Supplier:  LABCONCO
New Product
4' Logic Class II, Type A2 Biological Safety Cabinet with 8 sash opening, Service Fixture, UV Lamp, Vacu-Pass Portals and Base Stand, 230V, 50/60Hz, Schuko 1 * 1 items
Catalog # LBCP302481132
Supplier:  LABCONCO