Order Entry
Italy
ContactUsLinkComponent
Super Erecta(R) Post, special cut length, 69-3/8in.H, for stationary use, includes post cap and leveling bolt, stainless steel 1 * 1 items
Catalog # METO69CPS
New Product
Super Erecta(R) Post, special cut length, 69-3/8in.H, for stationary use, includes post cap and leveling bolt, stainless steel 1 * 1 items
Catalog # METO69CPS