Order Entry
Italy
ContactUsLinkComponent
Adapter USB-Wi-Fi/Bluetooth for the Mini Spray Dryer S-300 To connect the Mini Spray Dryer S-300 to Wi-Fi or generate a hot spot with the instrument. Connect to the App and control the machine remotely and download the reports. 1 * 1 items
Catalog # BUCH11072500
Supplier:  BUCHI
New Product
Adapter USB-Wi-Fi/Bluetooth for the Mini Spray Dryer S-300 To connect the Mini Spray Dryer S-300 to Wi-Fi or generate a hot spot with the instrument. Connect to the App and control the machine remotely and download the reports. 1 * 1 items
Catalog # BUCH11072500
Supplier:  BUCHI