Order Entry
Italy
ContactUsLinkComponent
witeg laboratory bottles, 250 ml, GL 45, borosilicate glass 3.3, clear, graduated, with witeg logo, ID code and certificate, autoclavable, ISO 4796-1, USP standard, with screw cap and pouring ring (blue, PP) Pack contains 10 Pcs 1 * 10 items
Catalog # WITG5.526.250.B
Supplier:  witeg Labortechnik
New Product
witeg laboratory bottles, 250 ml, GL 45, borosilicate glass 3.3, clear, graduated, with witeg logo, ID code and certificate, autoclavable, ISO 4796-1, USP standard, with screw cap and pouring ring (blue, PP) Pack contains 10 Pcs 1 * 10 items
Catalog # WITG5.526.250.B
Supplier:  witeg Labortechnik