Order Entry
Italy
ContactUsLinkComponent
1 component: Bromomethane [CAS:74-83-9] 5% w/w in Dimethyl sulphoxide CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 5 Ampoul
Catalog # CMPLRD0819511.5
New Product
1 component: Bromomethane [CAS:74-83-9] 5% w/w in Dimethyl sulphoxide CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 5 Ampoul
Catalog # CMPLRD0819511.5