Order Entry
Italy
Orders LinkContactUsLinkComponent
 

(4-Chlorophenyl)methyl acetate, tech

Bottle
Catalog # APOSOR32624-5G
Supplier:   Apollo Scientific
(4-Chlorophenyl)methyl acetate, tech
Catalog # APOSOR32624-5G
Supplier:  Apollo Scientific
CAS Number:  5406-33-7
(4-Chlorophenyl)methyl acetate, tech

Bottle

CAS Number:  5406-33-7
Price
3 030,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  p-Chlorobenzyl acetate, 4-Chlorobenzyl acetate
  • CAS Number:5406-33-7
  • Formula:C₉H₉ClO₂
  • MDL Number:MFCD00027406
  • Molecular Weight:184.62 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications