(4-Chlorophenyl)methyl acetate, tech
Bottle
Catalog # APOSOR32624-100MG
Supplier: Apollo Scientific
(4-Chlorophenyl)methyl acetate, tech
Catalog # APOSOR32624-100MG
Supplier: Apollo Scientific
CAS Number: 5406-33-7
Jump to:
Product Details & Documents
Synonyms: p-Chlorobenzyl acetate, 4-Chlorobenzyl acetate
- CAS Number:5406-33-7
- Formula:C₉H₉ClO₂
- MDL Number:MFCD00027406
- Molecular Weight:184.62 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



