Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Vitamin E 99%

Bottle
  APOSOR1015423-500G
 
New Product
Vitamin E 99%
  APOSOR1015423-500G
New Product
 :  59-02-9
Vitamin E 99%

Bottle

 :  59-02-9
 
215,00 €
   
1 * 500 g
 

Usage: Organic building block/ reagent.

 :  (+)-α-Tocopherol, Vitamin E Alcohol, (R)-2,5,7,8-Tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, α-Vitamin E, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E, D(+)-α-Tocopherol
  •  :59-02-9
  •  :C₂₉H₅₀O₂
  •  :430,71 g/mol
  •  :200-412-2
 

 

  • Pack type:
    Bottle