Order Entry
Italy
Orders LinkContactUsLinkComponent
Trypan Blue 80%

Trypan Blue 80%

Bottle
Catalog # APOSOR1015400-100G
Supplier:   Apollo Scientific
Trypan Blue 80%
Catalog # APOSOR1015400-100G
Supplier:  Apollo Scientific
CAS Number:  72-57-1
Trypan Blue 80%

Bottle

CAS Number:  72-57-1
Price
128,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
  • CAS Number:72-57-1
  • Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
  • MDL Number:MFCD00003969
  • Molecular Weight:960,81 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.