1,1'-Binaphthalene 98%
Bottle
Catalog # APOSOR80237-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: 1,1'-Binaphthalene
- CAS Number:604-53-5
- Formula:C₂₀H₁₄
- MDL Number:MFCD00041740
- Molecular Weight:254,33 g/mol
Specifications
Specifications
- Pack type:Bottle



