Vapour Pressure ASTM D5191: DVPE @ 51.6 kPa, Ptot @ 57.3 kPa 10 x 10 ml, 2,3-Dimethylbutane 1 * 100 mL
Catalog # PROOALK-CRMU-VP04K
Supplier: LGC Standards PROMOCHEM
New Product
Vapour Pressure ASTM D5191: DVPE @ 51.6 kPa, Ptot @ 57.3 kPa 10 x 10 ml, 2,3-Dimethylbutane 1 * 100 mL
Catalog # PROOALK-CRMU-VP04K
Supplier: LGC Standards PROMOCHEM
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF