Base/Neutrals Surrogate Standard Mixture - 3 components: 1000ug/ml each of 2-Fluorobiphenyl [CAS:321-60-8] Nitrobenzene D5 [CAS:4 165-60-0] p-Terphenyl D14 [CAS:1718-51-0] in Dichloromethane (10 x 1ml Ampoul) null
Catalog # CMPLF867791
Supplier: CUSTOM MADE CHEMICALS LAB