Order Entry
Italy
ContactUsLinkComponent
FEX 1000 EVO extraction unit with control electric Technical data: EX-protected for zone 22 Air flow rate 200 m /h in operating mode, measured at the end of 6 m hose Noise: 62 dBA at average power throughput Weight: 58 kg Powerful motor suitable for 1 * 1 items
  HAVE210766901
New Product
FEX 1000 EVO extraction unit with control electric Technical data: EX-protected for zone 22 Air flow rate 200 m /h in operating mode, measured at the end of 6 m hose Noise: 62 dBA at average power throughput Weight: 58 kg Powerful motor suitable for 1 * 1 items
  HAVE210766901