Order Entry
Italy
ContactUsLinkComponent
Injector module A 302/2 for optimum loading of laboratory glassware – can be equipped with 32 nozzles. 1 * 1 items
Catalog # MIEL12669210
Supplier:  MIELE
New Product
Injector module A 302/2 for optimum loading of laboratory glassware – can be equipped with 32 nozzles. 1 * 1 items
Catalog # MIEL12669210
Supplier:  MIELE