Heating Mantles WHM, for beakers 3000 ml, without integrated power controller, temperature range 450C, 550 W, power supply 230 V 1 * 1 items
Catalog # WITGDH.WHM12177
Supplier: witeg Labortechnik
New Product
Heating Mantles WHM, for beakers 3000 ml, without integrated power controller, temperature range 450C, 550 W, power supply 230 V 1 * 1 items
Catalog # WITGDH.WHM12177
Supplier: witeg Labortechnik
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Bottles, Narrow Neck, with Screw Cap
Supplier: RIXIUS