Accessories for mechanical overhead stirrer
Standard plate stand, with thread, plate (300×400 mm) with silicone rubber pad, stainless steel rod (Ø×L 23×1000 mm), All overhead stirrers and homogenizer
WITGDH.WOS501515
- Description:Standard plate stand, with thread, plate (300×400 mm) with silicone rubber pad, stainless steel rod (Ø×L 23×1000 mm)
- For:All overhead stirrers and homogenizer