Order Entry
Italy
ContactUsLinkComponent
Calibration temperature and humidity (MKF,MKFT) - including certificate, one measuring point in center of chamber at 25 °C / 60 % RH or at specified values 1 * 1 items
Catalog # BINDDL30-0330
Supplier:  Binder
New Product
Calibration temperature and humidity (MKF,MKFT) - including certificate, one measuring point in center of chamber at 25 °C / 60 % RH or at specified values 1 * 1 items
Catalog # BINDDL30-0330
Supplier:  Binder