Order Entry
Italy
ContactUsLinkComponent
Comfort rack for chest freezers for 7 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 298x140x140mm, del ivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24431B
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 7 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 298x140x140mm, del ivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24431B
Supplier:  TENAK