TriLink® 2'-Deoxyadenosine-5'-Triphosphate
Catalog # SOLUN-2506-25
Supplier: TriLink BioTechnologies
TriLink® 2'-Deoxyadenosine-5'-Triphosphate
Catalog # SOLUN-2506-25
Supplier: TriLink BioTechnologies
CAS Number:
Jump to:
Product Details & Documents
This nucleotide consists of an unmodified base, a deoxyribose, and a 5′ triphosphate group for synthesis of DNA molecules.
Unmodified dexoyribonucleotide for DNA synthesis
≥97% purity by AX-HPLC analysis
Provided as 100 mM in H₂O, pH 7,5
Grade: RUO
Deoxyadenosine-5′-triphosphate (dATP), deoxycytidine-5′-triphosphate (dCTP), deoxyguanosine-5′-triphosphate (dGTP), and deoxythymidine-5′-triphosphate (dTTP) are unmodified deoxyribonucleoside triphosphates, serving as essential building blocks of DNA molecules. Each of them consists of an unmodified base, a dexoyribose, and a 5′ triphosphate group.
Caution: Item will be delivered at or below −20 °C and should be stored at or below −20 °C.



