Order Entry
Italy
Orders LinkContactUsLinkComponent
TriLink® 2'-Deoxyadenosine-5'-Triphosphate

TriLink® 2'-Deoxyadenosine-5'-Triphosphate

Catalog # SOLUN-2506-100
TriLink® 2'-Deoxyadenosine-5'-Triphosphate
Catalog # SOLUN-2506-100
CAS Number:  
TriLink® 2'-Deoxyadenosine-5'-Triphosphate
TriLink® 2'-Deoxyadenosine-5'-Triphosphate
CAS Number:  
Price
206,00 €
Log in for availability
1 * 100 umol
Product Details & Documents

This nucleotide consists of an unmodified base, a deoxyribose, and a 5′ triphosphate group for synthesis of DNA molecules.

  • Unmodified dexoyribonucleotide for DNA synthesis

    ≥97% purity by AX-HPLC analysis

    Provided as 100 mM in H₂O, pH 7,5

    Grade: RUO

Deoxyadenosine-5′-triphosphate (dATP), deoxycytidine-5′-triphosphate (dCTP), deoxyguanosine-5′-triphosphate (dGTP), and deoxythymidine-5′-triphosphate (dTTP) are unmodified deoxyribonucleoside triphosphates, serving as essential building blocks of DNA molecules. Each of them consists of an unmodified base, a dexoyribose, and a 5′ triphosphate group.

Caution: Item will be delivered at or below −20 °C and should be stored at or below −20 °C.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.