Order Entry
Italy
ContactUsLinkComponent
Recommended for processing samples with Vivaflow tangential flow filtration (TFF) cassettes these Watson-Marlow pumps are ideal for ultrafiltration and diafiltration applications. 1 * 1 items
Catalog # SARTVF-APD0001-1
Supplier:  Sartorius
New Product
undefined
Recommended for processing samples with Vivaflow tangential flow filtration (TFF) cassettes these Watson-Marlow pumps are ideal for ultrafiltration and diafiltration applications. 1 * 1 items
Catalog # SARTVF-APD0001-1
Supplier:  Sartorius