COMPONENT-MPC 3/8IN INSERT 1 * 40 items
Catalog # SARTFBP101956
Supplier: Sartorius
New Product
Frequently Bought Together
VWR®, Beakers, Low Form, Heavy-Duty
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Tubing connectors, L-shaped
Supplier: KARTELL